Merck
  • Polyphenol-rich Rutgers Scarlet Lettuce improves glucose metabolism and liver lipid accumulation in diet-induced obese C57BL/6 mice.
Polyphenol-rich Rutgers Scarlet Lettuce improves glucose metabolism and liver lipid accumulation in diet-induced obese C57BL/6 mice.
Nutrition (Burbank, Los Angeles County, Calif.) (2014-07-06)
Diana M Cheng, Natalia Pogrebnyak, Peter Kuhn, Alexander Poulev, Carrie Waterman, Patricio Rojas-Silva, William D Johnson, Ilya Raskin
ABSTRACT
The aims of the following experiments were to characterize antidiabetic in vitro and in vivo activity of the polyphenol-rich aqueous extract of Rutgers Scarlet Lettuce (RSL). RSL extract (RSLE) and isolated compounds were evaluated for inhibitory effects on glucose production as well as tumor necrosis factor alpha-dependent inhibition of insulin activity in H4IIE rat hepatoma cells. Additionally, high-fat diet-induced obese mice were treated with RSLE (100 or 300 mg/kg), metformin (250 mg/kg), or vehicle (water) for 28 d by oral administration and insulin and oral glucose tolerance tests were conducted. Tissues were harvested at the end of the study and evaluated for biochemical and physiological improvements in metabolic syndrome conditions. A polyphenol-rich RSLE, containing chlorogenic acid, cyanidin malonyl-glucoside, and quercetin malonyl-glucoside, was produced by simple boiling water extraction at pH 2.0. In vitro, RSLE and chlorogenic acid demonstrated dose-dependent inhibition of glucose production. In vivo, RSLE treatment improved glucose metabolism measured by oral glucose tolerance tests, but not insulin tolerance tests. RSLE treated groups had a lower ratio of liver weight to body weight as well as decreased total liver lipids compared with the control group after 28 d of treatment. No significant differences in plasma glucose, insulin, cholesterol, and triglycerides were observed with RSLE-treated groups compared with vehicle control. RSLE demonstrated antidiabetic effects in vitro and in vivo and may improve metabolic syndrome conditions of fatty liver and glucose metabolism.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Chlorogenic acid, primary reference standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Quercetin 3-glucoside, analytical standard
Supelco
Quinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Isoquercitrin, primary reference standard