Merck
  • Water-soluble fullerene materials for bioapplications: photoinduced reactive oxygen species generation.
Water-soluble fullerene materials for bioapplications: photoinduced reactive oxygen species generation.
Faraday discussions (2014-12-04)
Yoko Yamakoshi, Safwan Aroua, Thach-Mien Duong Nguyen, Yuko Iwamoto, Tsuyoshi Ohnishi
ABSTRACT
The photoinduced reactive oxygen species (ROS) generation from several water-soluble fullerenes was examined. Macromolecular or small molecular water-soluble fullerene complexes/derivatives were prepared and their (1)O(2) and O(2)˙(-) generation abilities were evaluated by ESR spin-trapping methods. As a result, efficient (1)O(2) generation was detected from photoexcited C(60), not only in organic solvents, but also in aqueous media and especially from small molecule C(60)-carboxylic acid derivatives. Whereas efficient O(2)˙(-) generation was observed in the aqueous solution of the C(60)/γ-CD complex under photoirradiation.
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%