Merck
  • Oroxylin A inhibits hypoxia-induced invasion and migration of MCF-7 cells by suppressing the Notch pathway.
Oroxylin A inhibits hypoxia-induced invasion and migration of MCF-7 cells by suppressing the Notch pathway.
Anti-cancer drugs (2014-03-19)
Yao Cheng, Kai Zhao, Guojun Li, Jing Yao, Qinsheng Dai, Hui Hui, Zhiyu Li, Qinglong Guo, Na Lu
ABSTRACT
Tumor invasion and migration obstructs the treatment and prognosis of cancer. In this research, we investigated the effect of oroxylin A, a natural compound extracted from Scutellaria radix, the root of Scutellaria baicalensis, on inhibition of the invasion and migration of three different tumor cell lines: MCF-7, DU145, and HepG2. The results suggested that oroxylin A could inhibit hypoxia-induced migration and invasion of the three cell lines mentioned above. To study the detailed mechanisms, studies were carried out on MCF-7 cells and it was found that oroxylin A could regulate the expression of related markers in MCF-7 cells including E-cadherin, N-cadherin, and Vimentin. It was also found that oroxylin A inhibited the hypoxia-induced invasion and migration of MCF-7 cells by suppressing the Notch pathway. Oroxylin A inhibited N1ICD translocating to the nucleus and binding to epithelial-mesenchymal transition-related transcription factor Snail, thus suppressing the invasion and migration of MCF-7 cells. Therefore, oroxylin A is expected to be a promising candidate for antimetastasis treatment through suppression of the hypoxia-induced Notch pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human NOTCH1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Notch1
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder