Merck
  • An ultra-high-throughput spiral microfluidic biochip for the enrichment of circulating tumor cells.
An ultra-high-throughput spiral microfluidic biochip for the enrichment of circulating tumor cells.
The Analyst (2014-05-21)
Majid Ebrahimi Warkiani, Bee Luan Khoo, Daniel Shao-Weng Tan, Ali Asgar S Bhagat, Wan-Teck Lim, Yoon Sim Yap, Soo Chin Lee, Ross A Soo, Jongyoon Han, Chwee Teck Lim
ABSTRACT
The detection and characterization of rare circulating tumor cells (CTCs) from the blood of cancer patients can potentially provide critical insights into tumor biology and hold great promise for cancer management. The ability to collect a large number of viable CTCs for various downstream assays such as quantitative measurements of specific biomarkers or targeted somatic mutation analysis is increasingly important in medical oncology. Here, we present a simple yet reliable microfluidic device for the ultra-high-throughput, label-free, size-based isolation of CTCs from clinically relevant blood volumes. The fast processing time of the technique (7.5 mL blood in less than 10 min) and the ability to collect more CTCs from larger blood volumes lends itself to a broad range of potential genomic and transcriptomic applications. A critical advantage of this protocol is the ability to return all fractions of blood (i.e., plasma (centrifugation), CTCs and white blood cells (WBCs) (size-based sorting)) that can be utilized for diverse biomarker studies or time-sensitive molecular assays such as RT-PCR. The clinical use of this biochip was demonstrated by detecting CTCs from 100% (10/10) of blood samples collected from patients with advanced-stage metastatic breast and lung cancers. The CTC recovery rate ranged from 20 to 135 CTCs mL(-1) and obtained under high purity (of 1 CTC out of every 30-100 WBCs which gives ∼4 log depletion of WBCs). They were identified with immunofluorescence assays (pan-cytokeratin+/CD45-) and molecular probes such as HER2/neu.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard