Merck
  • Dorsal hippocampal microinjection of chlorpheniramine reverses the anxiolytic-like effects of l-histidine and impairs emotional memory in mice.
Dorsal hippocampal microinjection of chlorpheniramine reverses the anxiolytic-like effects of l-histidine and impairs emotional memory in mice.
Neuroscience letters (2014-12-20)
L Canto-de-Souza, D C Garção, F Romaguera, R Mattioli
ABSTRACT
Several findings have pointed to the role of histaminergic neurotransmission in the modulation of anxiety-like behaviors and emotional memory. The elevated plus-maze (EPM) test has been widely used to investigate the process of anxiety and also has been used to investigate the process of learning and memory. Visual cues are relevant to the formation of spatial maps, and as the hippocampus is involved in this task, experiment 1 explored this issue. Experiment 2 investigated the effects of intraperitoneal (i.p.) injections of l-histidine (LH, a precursor of histamine) and of intra-dorsal hippocampus (intra-DH) injections of chlorpheniramine (CPA, an H1 receptor antagonist) on anxiety and emotional memory in mice re-exposed to the EPM. Mice received saline (SAL) or LH i.p. and SAL or CPA (0.016, 0.052, and 0.16 nmol/0.1 μl) intra-DH prior to Trial 1 (T1) and Trial 2 (T2). No significant changes were observed in the number of enclosed-arm entries (EAE) in T1, an EPM index of general exploratory activity. LH had an anxiolytic-like effect that was reversed by intra-DH injections of CPA. T2 versus T1 analysis revealed that only the lower dose of CPA resulted in impaired emotional memory. Combined injections of LH and CPA revealed that higher doses of CPA impair emotional memory. Taken together, these results suggest that LH and H1 receptors present in the dorsal hippocampus are involved in anxiety-related behaviors and emotional memory in mice submitted to EPM.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Chlorpheniramine maleate, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard