Merck
  • Zinc sulfide nanoparticles selectively induce cytotoxic and genotoxic effects on leukemic cells: involvement of reactive oxygen species and tumor necrosis factor alpha.
Zinc sulfide nanoparticles selectively induce cytotoxic and genotoxic effects on leukemic cells: involvement of reactive oxygen species and tumor necrosis factor alpha.
Journal of applied toxicology : JAT (2014-01-31)
Sandeep Kumar Dash, Totan Ghosh, Soumyabrata Roy, Sourav Chattopadhyay, Debasis Das
ABSTRACT
The aim of the present study was to develop zinc sulfide nanoparticles (ZnS NPs) and to study their cytotoxicity against the KG-1A (human acute myeloid leukemia) cell line. ZnS NPs were synthesized using the pyrolytic method and characterized by X-ray diffraction, dynamic light scattering, surface zeta potential, scanning electron microscopy and atomic force microscopy. Cell viability study and flow cytometric analysis confirmed the potent cytotoxic effects of ZnS NPs on cancer cells in a dose-dependent fashion. Successful uptakes of ZnS NPs by leukemic cells were confirmed by phase contrast fluorescence microscopy. pH-dependent dissolution of ZnS NPs was done using atomic absorption microscopy to understand the cell-specific internalization of Zn(+) . This internalization of NPs facilitated the generation of excess reactive oxygen species (ROS), followed by tumor necrosis factor alpha (TNF-α) secretion which caused severe DNA damage as observed in the comet assay and altered the mitochondrial membrane potential (MMP) in leukemic cells. Surprisingly ZnS NPs had no toxic effects on normal lymphocytes at doses up to 50 µg ml(-1) . Pre-treatment with ROS and TNF-α inhibitor confirmed that these nanoparticles were able to kill leukemic cells by generating an excess amount of ROS and thereby initiated TNF-α mediated apoptosis pathway. These findings clarify the mechanism with which ZnS NPs induced anticancer activities in vitro. To elicit its utilities and its application to cancer treatment in vivo is under investigation.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Sigma-Aldrich
2-Benzoylpyridine, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione oxidized, lyophilized powder
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)
SAFC
L-Glutathione oxidized
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, Dye content 90 %