Merck
  • In vitro evaluation of inorganic mercury and methylmercury effects on the intestinal epithelium permeability.
In vitro evaluation of inorganic mercury and methylmercury effects on the intestinal epithelium permeability.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-12-03)
M Vázquez, D Vélez, V Devesa
ABSTRACT
The mercurial forms [inorganic divalent mercury, Hg(II) and methylmercury, CH3Hg] produce neurological and immune effects as well as hematological and renal alterations. The main route of exposure is through the diet. Consequently, the gastrointestinal mucosa is exposed to these mercurial forms, though the potential toxic effects upon the mucosa are not clear. The present study evaluates the toxicity of Hg(II) and CH3Hg (0.1-2 mg/L) in an intestinal epithelium model using the differentiated and undifferentiated human Caco-2 cell line.The experiments made show the mercurial forms generate reactive oxygen and/or nitrogen species and a significant decrease in glutathione contents. This redox imbalance could be the cause of the lipid peroxidation observed after short exposure times. Such conditions of stress lead to a modulation of stress proteins, intercellular junction proteins and tumor necrosis factor-alpha expression and to a redistribution of F-actin and ZO1 protein in the intestinal monolayer. The abovementioned effects may be the cause of the increase in permeability in the differentiated cells observed at concentrations similar to those found in food products (0.5-1 mg/L). The increase in permeability could produce an impairment of the barrier function of the intestinal epithelium.
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
SAFC
HEPES
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Sulfosalicylic acid dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
5-Sulfosalicylic acid dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
5-Sulfosalicylic acid dihydrate, suitable for electrophoresis, ≥99%
Sigma-Aldrich
5-Sulfosalicylic acid dihydrate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
5-Sulfosalicylic acid dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder