Merck
  • Development of a second generation palladium-catalyzed cycloalkenylation and its application to bioactive natural product synthesis.
Development of a second generation palladium-catalyzed cycloalkenylation and its application to bioactive natural product synthesis.
Natural product communications (2013-08-29)
Masahiro Toyota
ABSTRACT
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This protocol has been applied to the total syntheses of cis-195A, trans-195A, boonein, scholareins A, C, D, and alpha-skytanthine.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 25x25mm, thickness 0.50mm, as rolled, 99.99+%
Palladium, foil, 10mm disks, thickness 0.008mm, 99.95%
Palladium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.02mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.0125mm, 99.95%
Palladium, foil, 10x10mm, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 6mm disks, thickness 0.0125mm, 99.95%
Palladium, foil, 4mm disks, thickness 0.008mm, 99.95%
Palladium, foil, 10mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%