Skip to Content
Merck
All Photos(1)

Documents

348643

Sigma-Aldrich

Palladium

foil, thickness 0.25 mm, 99.98% trace metals basis

Synonym(s):

Palladium (powder), Palladium black, Palladium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98% trace metals basis

form

foil

resistivity

9.96 μΩ-cm, 20°C

thickness

0.25 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Quantity

1.9 g = 25 × 25 mm; 7.6 g = 50 × 50 mm

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Bin Kim et al.
Journal of nanoscience and nanotechnology, 13(1), 517-522 (2013-05-08)
In order to form flexible printed circuits through inkjet printing technique, the Pd(ll) catalyst ink was printed on the surface of polyimide film modified with KOH solution and then reduced with NaBH4 solution to extract the Pd(O) catalyst nuclei. The
Xiaohu Xia et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6669-6673 (2013-04-10)
Controlling the shape or morphology of metal nanocrystals is central to the realization of their many applications in catalysis, plasmonics, and electronics. In one of the approaches, the metal nanocrystals are grown from seeds of certain crystallinity through the addition
Hyunsu Kim et al.
Journal of nanoscience and nanotechnology, 13(1), 533-536 (2013-05-08)
ZnSnO3 one-dimensional (1D) strutures were synthesized by using an evaporation technique. The morphology, crystal structure, and enhanced sensing properties of the ZnSnO3 structures functionalized with Pd to CO gas at 300 degrees C were investigated. The diameters of the 1D
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service