Merck
  • Water in Photosystem II: structural, functional and mechanistic considerations.
Water in Photosystem II: structural, functional and mechanistic considerations.
Biochimica et biophysica acta (2013-08-28)
Katrin Linke, Felix M Ho
ABSTRACT
Water is clearly important for the functioning of Photosystem II (PSII). Apart from being the very substrate that needs to be transported in this water oxidation enzyme, water is also vital for the transport of protons to and from the catalytic center as well as other important co-factors and key residues in the enzyme. The latest crystal structural data of PSII have enabled detailed analyses of the location and possible function of water molecules in the enzyme. Significant progress has also been made recently in the investigation of channels and pathways through the protein complex. Through these studies, the mechanistic significance of water for PSII is becoming increasingly clear. An overview and discussion of key aspects of the current research on water in PSII is presented here. The role of water in three other systems (aquaporin, bacteriorhodopsin and cytochrome P450) is also outlined to illustrate further points concerning the central significance that water can have, and potential applications of these ideas for continued research on PSII. It is advocated that water be seen as an integral part of the protein and far from a mere solvent.
Product Description
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, for inorganic trace analysis, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture