Merck
  • Iodocyclization, followed by palladium-catalyzed coupling: a versatile strategy for heterocyclic library construction.
Iodocyclization, followed by palladium-catalyzed coupling: a versatile strategy for heterocyclic library construction.
Combinatorial chemistry & high throughput screening (2012-01-26)
Anton V Dubrovskiy, Nataliya A Markina, Richard C Larock
ABSTRACT
The iodocyclization of functionally-substituted alkynes provides an excellent way to prepare a wide range of iodoheterocycles, which can then be readily elaborated through palladium-catalyzed Suzuki-Miyaura, Sonogashira, Heck, Hartwig-Buchwald, and carbonylation processes into libraries of medicinally relevant heterocycles. The synthesis of libraries of indoles, benzofurans, benzothiophenes, isocoumarins and pyrones, cyclic imidates, isoxazoles, furans using this approach is reviewed. This technology is very versatile, proceeds under mild reaction conditions in high yields, and tolerates considerable functionality.
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Palladium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, light tested, 25x25mm, thickness 0.004mm, 99.95%
Palladium, wire reel, 0.5m, diameter 0.125mm, hard, 99.95%
Palladium, wire reel, 2m, diameter 0.025mm, as drawn, 99.9%
Palladium, tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, wire reel, 0.1m, diameter 1.0mm, hard, 99.95%
Palladium, wire reel, 0.5m, diameter 0.25mm, hard, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.95%
Palladium, foil, not light tested, 150x150mm, thickness 0.03mm, as rolled, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.003mm, 99.95%