Merck
  • Preparation and characterization of fucoidan-chitosan nanospheres by the sonification method.
Preparation and characterization of fucoidan-chitosan nanospheres by the sonification method.
Journal of nanoscience and nanotechnology (2014-04-17)
Yuangang Liu, Wenbin Yao, Shibin Wang, Geng Di, Qiyao Zheng, Aizheng Chen
ABSTRACT
Fucoidan was chosen a substitute for alginate in chitosan-based polyelectrolytes or polyiones because it is negatively charged and possesses pharmacological properties. The fucoidan-chitosan nanospheres were prepared by the sonification method, and different formulation variables were investigated to get regular and round microspheres. Single factor and orthogonal experiments were carried out in this process, and nanospheres were finally prepared successfully. Stability tests revealed that some of the spheres were stable over one week. Combined with the drug loading/release profiles when using bovine serum albumin and capecitabine as model drugs for peptide and small molecule drug respectively, it can be concluded that the nanospheres may have potential applications in drug delivery.
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Fucoidan from Macrocystis pyrifera, ≥85%
Sigma-Aldrich
Fucoidan from Undaria pinnatifida, ≥95%
Sigma-Aldrich
Fucoidan from Fucus vesiculosus, ≥95%
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Fucoidan from Fucus vesiculosus, Crude