Skip to Content
Merck
All Photos(1)

Key Documents

F8315

Sigma-Aldrich

Fucoidan from Undaria pinnatifida

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

algae (Undaria pinnatifida)

Quality Level

Assay

≥95%

form

powder

color

white to off-white

useful pH range

3.0 -7.0

solubility

water: 10 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

room temp

General description

Fucoidan has a complex chemical composition. It contains mannose, galactose, glucose, xylose and other monosaccharides. It is also composed of uronic acids, acetyl groups and protein.[1]
Fucoidan is a fucose-rich, sulfated polysaccharide isolated from several species of brown seaweed.

Application

Fucoidan from Undaria pinnatifida has been used to assess its antimicrobial potential in vitro.[2]

Biochem/physiol Actions

Studies of fucoidan have found it to have a broad variety of biological activities including antitumor and antiangogenic activities, immunomodulatory, anti-inflammatory, anticoagulant, antithrombotic, and antioxidant activities. Fucoidan induces macrophage activation which subsequently activates mitogen activated protein kinases (MAPKs) and results in natural killer cell proliferation. Fucoidan acts as a nonselective selectin blocker and has been shown to induce apoptosis and suppress angiogenesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fucoidan: structure and bioactivity
Li B, et al.
Molecules (Basel), 13(8), 1671-1695 (2008)
Arumugam Ponnan et al.
Environmental science and pollution research international, 27(25), 31760-31766 (2020-06-07)
Fucoidan is a unique bioactive and dietary polymer enriched mainly in the cell wall matrix of the brown seaweeds. This present study was intended to reveal the antigenotoxicity effect of fucoidan on 4-nitroquinolin-1-oxide (4-NQO) induced genetics damage and apoptosis in
Anna Pielesz et al.
The Analyst, 140(13), 4599-4607 (2015-06-02)
Differential scanning calorimetry (DSC) and thermogravimetric (TGA) investigations, acetate electrophoresis (CAE), Fourier-transform infrared spectrometry (FTIR), scanning electron microscopy (SEM) analysis and microbiological procedures were all carried out after heating the samples to a temperature sufficient for simulating a burn incident.
Xiang Li et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(8), 1661-1667 (2014-06-07)
Nuclear imaging of active plaques still remains challenging. Advanced atherosclerotic plaques have a strong expression of P-selectin by the endothelium overlying active atherosclerotic plaques, but not on the endothelium overlying inactive fibrous plaques. We proposed a new approach for noninvasive
Wentong Cui et al.
Laboratory investigation; a journal of technical methods and pathology, 94(4), 382-393 (2014-03-13)
Endothelial dysfunction, characterized by impairment of endothelial nitric oxide synthase (eNOS) and nitric oxide (NO) bioavailability, has been implicated in diabetic cardiovascular pathogenesis. In this study, low-molecular-weight fucoidan (LMWF), which has multiple biological activities including anti-inflammatory and anti-oxidative properties, was

Questions

  1. Could you please share a reference to the extraction and purification method used for this sample? I have it as a standard in my exxperiment and would like to get insights into it. Thank you

    1 answer
    1. The extraction and purification methods for this product are considered proprietary information.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service