Merck
  • In situ intracellular calcium oscillations in osteocytes in intact mouse long bones under dynamic mechanical loading.
In situ intracellular calcium oscillations in osteocytes in intact mouse long bones under dynamic mechanical loading.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-12-19)
Da Jing, Andrew D Baik, X Lucas Lu, Bin Zhou, Xiaohan Lai, Liyun Wang, Erping Luo, X Edward Guo
ABSTRACT
Osteocytes have been hypothesized to be the major mechanosensors in bone. How in situ osteocytes respond to mechanical stimuli is still unclear because of technical difficulties. In vitro studies have shown that osteocytes exhibited unique calcium (Ca(2+)) oscillations to fluid shear. However, whether this mechanotransduction phenomenon holds for in situ osteocytes embedded within a mineralized bone matrix under dynamic loading remains unknown. Using a novel synchronized loading/imaging technique, we successfully visualized in real time and quantified Ca(2+) responses in osteocytes and bone surface cells in situ under controlled dynamic loading on intact mouse tibia. The resultant fluid-induced shear stress on the osteocyte in the lacunocanalicular system (LCS) was also quantified. Osteocytes, but not surface cells, displayed repetitive Ca(2+) spikes in response to dynamic loading, with spike frequency and magnitude dependent on load magnitude, tissue strain, and shear stress in the LCS. The Ca(2+) oscillations were significantly reduced by endoplasmic reticulum (ER) depletion and P2 purinergic receptor (P2R)/phospholipase C (PLC) inhibition. This study provides direct evidence that osteocytes respond to in situ mechanical loading by Ca(2+) oscillations, which are dependent on the P2R/PLC/inositol trisphosphate/ER pathway. This study develops a novel approach in skeletal mechanobiology and also advances our fundamental knowledge of bone mechanotransduction.
Product Description
Neomycin sulfate, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
USP
Neomycin sulfate, United States Pharmacopeia (USP) Reference Standard
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard