Merck
  • In older men an optimal plasma testosterone is associated with reduced all-cause mortality and higher dihydrotestosterone with reduced ischemic heart disease mortality, while estradiol levels do not predict mortality.
In older men an optimal plasma testosterone is associated with reduced all-cause mortality and higher dihydrotestosterone with reduced ischemic heart disease mortality, while estradiol levels do not predict mortality.
The Journal of clinical endocrinology and metabolism (2013-11-22)
Bu B Yeap, Helman Alfonso, S A Paul Chubb, David J Handelsman, Graeme J Hankey, Osvaldo P Almeida, Jonathan Golledge, Paul E Norman, Leon Flicker
ABSTRACT
Testosterone (T) levels decline with age and lower T has been associated with increased mortality in aging men. However, the associations of its metabolites, dihydrotestosterone (DHT) and estradiol (E2), with mortality are poorly defined. We assessed associations of T, DHT, and E2 with all-cause and ischemic heart disease (IHD) mortality in older men. Participants were community-dwelling men aged 70 to 89 years who were residing in Perth, Western Australia. Plasma total T, DHT, and E2 were assayed using liquid chromatography-tandem mass spectrometry in early morning samples collected in 2001 to 2004 from 3690 men. Deaths to December 2010 were ascertained by data linkage. There were 974 deaths (26.4%), including 325 of IHD. Men who died had lower baseline T (12.8±5.1 vs 13.2±4.8 nmol/L [mean±SD], P=.013), DHT (1.4±0.7 vs 1.5±0.7 nmol/L, P=.002), and E2 (71.6±29.3 vs 74.0±29.0 pmol/L, P=.022). After allowance for other risk factors, T and DHT were associated with all-cause mortality (T: quartile [Q] Q2:Q1, adjusted hazard ratio [HR]=0.82, P=.033; Q3:Q1, HR=0.78, P=.010; Q4:Q1, HR=0.86, P>.05; DHT: Q3:Q1, HR=0.76, P=.003; Q4:Q1, HR=0.84, P>.05). Higher DHT was associated with lower IHD mortality (Q3:Q1, HR=0.58, P=.002; Q4:Q1, HR=0.69, P=.026). E2 was not associated with either all-cause or IHD mortality. Optimal androgen levels are a biomarker for survival because older men with midrange levels of T and DHT had the lowest death rates from any cause, whereas those with higher DHT had lower IHD mortality. Further investigations of the biological basis for these associations including randomized trials of T supplementation are needed.
Product Description
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®