Merck
Immunity, inflammation and cancer: a leading role for adenosine.
Nature reviews. Cancer (2013-11-15)
Luca Antonioli, Corrado Blandizzi, Pál Pacher, György Haskó
ABSTRACT
Cancer is a complex disease that is dictated by both cancer cell-intrinsic and cell-extrinsic processes. Adenosine is an ancient extracellular signalling molecule that can regulate almost all aspects of tissue function. As such, several studies have recently highlighted a crucial role for adenosine signalling in regulating the various aspects of cell-intrinsic and cell-extrinsic processes of cancer development. This Review critically discusses the role of adenosine and its receptors in regulating the complex interplay among immune, inflammatory, endothelial and cancer cells during the course of neoplastic disease.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine