Merck
  • Preparation, characterization and tableting of cilnidipine solid dispersions.
Preparation, characterization and tableting of cilnidipine solid dispersions.
Pakistan journal of pharmaceutical sciences (2013-04-30)
Liandong Hu, Weihua Song, Feng Niu, Kuiliang Jiao, Zheng Jia
ABSTRACT
Solid dispersion technique has been developed many years for improving solubility of water-insoluble drugs, aiming to achieve a better oral bioavailability. However, this technique exhibits many inconveniences when used for large-scale tableting procedures. The objective of current research work was to develop cilnidipine solid dispersions (SDs) to improve the dissolution behaviors of this water-insoluble drug. Moreover, an innovative granulation method was designed to simplify the traditional tableting technology used in solid dispersion technique. Three different kinds of polymers, polyethylene glycol (PEG), polyvinylpyrrolidone (PVP) and poloxamer, were used as carriers to prepare solid dispersions. The interactions in the solid state were characterized by differential scanning calorimetry (DSC), powder X-ray diffraction (PXRD) and FT-IR spectroscopy. The designed granulation method was employed to prepare solid dispersion tablets and the formulation was optimized through investigating the dissolution behaviors. The results indicated PEG solid dispersion showed the best effect both on physical characterizations and dissolution studies. Furthermore, all type of solid dispersions significantly improved the dissolution rates when compared to pure drug and its corresponding physical mixture (PM). The solid dispersion tablets prepared in simplified tableting method exhibited better operability, stability and dissolution behavior than the tablets prepared in traditional ways, which brought more opportunities to solid dispersion technique for industrial production.
Sigma-Aldrich
Kollidon® 25
Povidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Cilnidipine, ≥98% (HPLC), powder
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 90