Skip to Content
Merck

Skip To

P0930

Polyvinylpyrrolidone

powder, BioXtra, suitable for mouse embryo cell culture

Synonym(s):

PVP, Polyvidone, Povidone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
NACRES:
NA.75
PubChem Substance ID:
UNSPSC Code:
12352207
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

form

powder

technique(s)

cell culture | embryo: suitable

viscosity number

28-32(lit.)

solubility

H2O: 100 mg/mL

application(s)

agriculture

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

General description

Polyvinylpyrrolidone (PVP)/polyvidone is used in cosmetics, medical and hair care products. Povidone iodine, a compound of polyvinylpyrrolidone and iodine can be used as an antiseptic and antibacterial agent. Dry form of PVP is available as a light flaky hygroscopic powder, which can absorb up to 40% of water by its weight.

Application

Polyvinylpyrrolidone has been used as a component in phosphate buffer saline (PBS) for immunostaining. It has also been used as a component in phosphate buffer saline (PBS) to wash parthenogenetic embryos for immunocytochemical analysis.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P5288856568PVP10
form

powder

form

powder

form

powder

form

powder

technique(s)

cell culture | embryo: suitable

technique(s)

-

technique(s)

-

technique(s)

-

application(s)

agriculture

application(s)

-

application(s)

-

application(s)

-

product line

BioXtra

product line

-

product line

-

product line

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

H2O: 100 mg/mL

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Polyvinylpyrrolidone


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. PVP can be dissolve in 1- pentanol or 1- butanol?

    1 answer
    1. This product is tested for solubility in water but is essentially identical to PVP K40. Various sources state that this material is soluble in methanol, ethanol, alcohol, chloroform,
      glycerol, and acetic acid. This information has not been validated. The material is likely to be soluble in pentanol and butanol, but this would have to be determined experimentally by the end user.

      Helpful?

  2. What is molecular average weight of PVP P0930?

    1 answer
    1. The average molecular weight of this product is 40 kDa. The range and distribution have not been established.

      Helpful?

Reviews

No rating value

Active Filters