Merck
  • Microwave accelerated selective Soxhlet extraction for the determination of organophosphorus and carbamate pesticides in ginseng with gas chromatography/mass spectrometry.
Microwave accelerated selective Soxhlet extraction for the determination of organophosphorus and carbamate pesticides in ginseng with gas chromatography/mass spectrometry.
Analytical chemistry (2012-06-13)
Ting Zhou, Xiaohua Xiao, Gongke Li
ABSTRACT
Microwave accelerated selective Soxhlet extraction (MA-SSE), a novel selective extraction technique, was investigated in this study. A Soxhlet extraction system containing a glass filter was designed as an extractor. During the procedure of MA-SSE, both the target analytes and the interfering components were extracted from the sample into the extraction solvent enhanced by microwave irradiation. After the solvent flowed though the sorbent, the interfering components were adsorbed by the sorbent, and the target analytes remaining in the solvent were collected in the extraction bottle. No cleanup or filtration was required after extraction. The efficiency of the MA-SSE approach was demonstrated in the determination of organophosphorus and carbamate pesticide residues in ginseng by gas chromatography/mass spectrometry (GC/MS). Under the optimized conditions, low limits of detection (0.050-0.50 μg/kg) were obtained. The recoveries were in the range of 72.0-110.1% with relative standard deviations less than 7.1%. Because of the effect of microwave irradiation, MA-SSE showed significant advantage compared with other extraction techniques. The sorbent used in this study showed good cleanup ability. The mechanism of MA-SSE was demonstrated to be based on the rupture of the cell walls according to the structural changes of ginseng samples. On the basis of the results, MA-SSE as a simple and effective sample preparation technique for the analysis of pesticide residues in complex matrixes shows great promise.
Product Description
Supelco
Florisil® Adsorbent for Chromatography, 60-100 mesh
Supelco
Florisil® Adsorbent for Chromatography, 30-60 mesh
Sigma-Aldrich
Florisil®, 100-200 mesh
Supelco
Florisil®, PR grade, 60-100 mesh, coarse powder
Supelco
Florisil®, 100-200 mesh, fine powder
Supelco
Florisil®, for the determination of hydrocarbon acc. to ISO 9377-2
Supelco
Florisil® PR, PR grade (analysis acc. to FDA)
Supelco
Florisil®, <200 mesh, fine powder