Merck
  • Chemiluminescence determination of terbutaline sulfate in bovine urine and pharmaceutical preparations based on enhancement of the 2-phenyl-4, 5-di (2-furyl) imidazole-potassium ferricyanide system.
Chemiluminescence determination of terbutaline sulfate in bovine urine and pharmaceutical preparations based on enhancement of the 2-phenyl-4, 5-di (2-furyl) imidazole-potassium ferricyanide system.
Journal of pharmaceutical and biomedical analysis (2011-10-15)
Lu Han, Yumin Zhang, Jing Kang, Jieli Tang, Yihua Zhang
ABSTRACT
In this paper, a novel chemiluminescence (CL) system, 2-phenyl-4, 5-di (2-furyl) imidazole (PDFI)-potassium ferricyanide, for the determination of terbutaline sulfate was described. The method was based on enhancement of CL emission of PDFI-potassium ferricyanide system in the presence of terbutaline sulfate. Under the optimum conditions, the enhanced chemiluminescence intensity is linearly related to the concentration of terbutaline sulfate. The proposed method has been successfully applied to the determination of terbutaline sulfate in bovine urine and pharmaceutical preparations with satisfactory results. Furthermore, the possible mechanism of chemiluminescence reaction was also discussed briefly.
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
Sigma-Aldrich
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%