Skip to Content
Merck
All Photos(2)

Key Documents

455946

Sigma-Aldrich

Potassium hexacyanoferrate(III)

99.98% trace metals basis

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.98% trace metals basis
form:
crystalline

Quality Level

Assay

99.98% trace metals basis

form

crystalline

impurities

≤250.0 ppm Trace Metal Analysis

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Potassium ferricyanide dopant may be used to co-dope Li4Ti5O12 (LTO) with K and Fe to enhance its electrochemical performance during the use of LTO as an anode for Li-ion batteries.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using potassium ferricyanide as a dopant to prepare K and Fe co-doped Li4Ti5O12
Ding K, et al.
Ceramics International, 42.16, 19187-19194 (2016)
Effects of cathodic electron acceptors and potassium ferricyanide concentrations on the performance of microbial fuel cell
Wei L, et al.
International Journal of Hydrogen Energy, 37.17, 12980-12986 (2012)
Holger Schulze et al.
Sensors (Basel, Switzerland), 21(5) (2021-04-04)
Rapid point of care tests for bacterial infection diagnosis are of great importance to reduce the misuse of antibiotics and burden of antimicrobial resistance. Here, we have successfully combined a new class of non-biological binder molecules with electrochemical impedance spectroscopy
Sveinung Lillehaug et al.
Scientific data, 6, 190028-190028 (2019-02-27)
The spatial pattern of transgene expression in tetracycline-controlled mouse models is governed primarily by the driver line used to introduce the tetracycline-controlled transactivator (tTA). Detailed maps showing where each tTA driver activates expression are therefore essential for designing and using
Akhtar Hayat et al.
Analytica chimica acta, 724, 92-97 (2012-04-10)
Okadaic acid (OA), a lipophilic phycotoxin is mainly produced by toxigenic dinoflagellates. The need to develop high performing methods for OA analysis able to improve the traditional ones is evident. In this work, a novel experimental methodology for label-free detection

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service