Merck
  • Fluorescent one-dimensional nanostructures from a group of uniform materials based on organic salts.
Fluorescent one-dimensional nanostructures from a group of uniform materials based on organic salts.
Chemical communications (Cambridge, England) (2011-06-16)
Sergio L de Rooy, Bilal El-Zahab, Min Li, Susmita Das, Ellen Broering, Lin Chandler, Isiah M Warner
ABSTRACT
Herein we report the synthesis of a fluorescent organic salt through anion exchange and the subsequent fabrication of 1D-nanostructures via a facile templating method.
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)
Sigma-Aldrich
Potassium tetraphenylborate, 97%
Sigma-Aldrich
Ammonium tetraphenylborate, 99%