Skip to Content
Merck
All Photos(1)

Key Documents

466689

Sigma-Aldrich

Potassium tetraphenylborate

97%

Synonym(s):

Potassium tetrakis(phenyl)borate, Potassium tetraphenylboride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4BK
CAS Number:
Molecular Weight:
358.32
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: boron
reagent type: catalyst

mp

>300 °C (lit.)

SMILES string

[K+].c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.K/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;/q-1;+1

InChI key

MAMCUZQNCMYPCY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dejan Stekovic et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(16), 4166-4174 (2018-12-28)
The ability to tune the physical properties of bistable organic functional materials by means of chemistry can facilitate their development for molecular electronic switching components. The butylamine-containing biphenalenyl boron neutral radical, [Bu]2 B, crystalline compound has recently attracted significant attention
Hiroyuki Hayashi et al.
The Journal of organic chemistry, 73(22), 8683-8693 (2008-10-16)
1-Ethyl-2-(1-oxy-3-oxo-4,4,5,5-tetramethylimidazolin-2-yl)-3-methylimidazolium bromide, [EMINN](+)[Br](-), carrying nitronylnitroxide (NN) in the cation unit, was prepared as a parent molecule and converted to seven salts, [EMINN](+)[X](-) (X = I, TFSI (bis(trifluoromethanesulfonyl)imide), BPh4 (tetraphenylborate), [EMINN](+)(1-3)[BA(1-3)]((1-3)-); BA1 (benzoic acid), BA2 (terephthalic acid), and BA3 (trimesic acid)
Beata Paczosa-Bator et al.
Talanta, 81(3), 1003-1009 (2010-03-20)
This paper focuses on the quantitative determination of the loss of the components from plastic membranes of ion-selective electrodes (ISEs) during contact with aqueous bathing solutions. The leaching processes, which affect the ISE responses, are rarely characterized by independent methods.
Mirela Samardžić et al.
Talanta, 83(3), 789-794 (2010-12-15)
A sensitive potentiometric surfactant sensor based on a highly lipophilic 1,3-didecyl-2-methyl-imidazolium cation and a tetraphenylborate (TPB) antagonist ion was used as the end-point detector in ion-pair potentiometric surfactant titrations using sodium TPB as a titrant. Several analytical and technical grade
Sergio L de Rooy et al.
Chemical communications (Cambridge, England), 47(31), 8916-8918 (2011-06-16)
Herein we report the synthesis of a fluorescent organic salt through anion exchange and the subsequent fabrication of 1D-nanostructures via a facile templating method.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service