Merck
  • Bismuth coordination chemistry with allyl, alkoxide, aryloxide, and tetraphenylborate ligands and the {[2,6-(Me2NCH2)2C6H3]2Bi}+ cation.
Bismuth coordination chemistry with allyl, alkoxide, aryloxide, and tetraphenylborate ligands and the {[2,6-(Me2NCH2)2C6H3]2Bi}+ cation.
Inorganic chemistry (2011-01-05)
Ian J Casely, Joseph W Ziller, Bruce J Mincher, William J Evans
ABSTRACT
A series of bis(aryl) bismuth compounds containing (N,C,N)-pincer ligands, [2,6-(Me(2)NCH(2))(2)C(6)H(3)](-) (Ar'), have been synthesized and structurally characterized to compare the coordination chemistry of Bi(3+) with similarly sized lanthanide ions, Ln(3+). Treatment of Ar'(2)BiCl, 1, with ClMg(CH(2)CH═CH(2)) affords the allyl complex Ar'(2)Bi(η(1)-CH(2)CH═CH(2)), 2, in which only one allyl carbon atom coordinates to bismuth. Complex 1 reacts with KO(t)Bu and KOC(6)H(3)Me(2)-2,6 to yield the alkoxide Ar'(2)Bi(O(t)Bu), 3, and aryloxide Ar'(2)Bi(OC(6)H(3)Me(2)-2,6), 4, respectively, but the analogous reaction with the larger KOC(6)H(3)(t)Bu(2)-2,6 forms [Ar'(2)Bi][OC(6)H(3)(t)Bu(2)-2,6], 6, in which the aryloxide ligand acts as an outer sphere anion. Chloride is removed from 1 by NaBPh(4) to form [Ar'(2)Bi][BPh(4)], 5, which crystallizes from THF in an unsolvated form with tetraphenylborate as an outer sphere counteranion.
Sigma-Aldrich
Potassium tetraphenylborate, 97%
Sigma-Aldrich
Ammonium tetraphenylborate, 99%
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)