Merck
  • Enhanced stability and decolorization of Coomassie Brilliant Blue R-250 by dextran aldehyde-modified horseradish peroxidase.
Enhanced stability and decolorization of Coomassie Brilliant Blue R-250 by dextran aldehyde-modified horseradish peroxidase.
Artificial cells, blood substitutes, and immobilization biotechnology (2010-12-02)
Melda Altikatoglu, Mithat Celebi
ABSTRACT
Horseradish peroxidase (EC 1.11.1.7) was chemically modified by periodate-activated dextran. The activities of free and modified enzyme against organic-aqueous interface and some chemicals were determined. Modified HRP remained fully active in the presence of organic solvent for 4 h. However, the unmodified enzyme lost 50% of its activity within the first 2 h. The effects of possible inhibitors on enzyme activity were investigated. In addition, Coomassie Brilliant Blue R-250 was efficiently decolorized using the free and modified HRP. After 5 minutes of treatment, the color removal of dye was 80-90%. Modified HRP showed effective performance compared to free HRP.
Sigma-Aldrich
Brilliant Blue R, 250, for microscopy
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution
Sigma-Aldrich
Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade
Sigma-Aldrich
Brilliant Blue R, pure