Skip to Content
Merck
All Photos(1)

Key Documents

B8647

Sigma-Aldrich

Brilliant Blue R Concentrate

methanol solution

Synonym(s):

Brilliant Blue R, Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution

form

methanol solution

Quality Level

color

dark blue

suitability

suitable for SDS-PAGE

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant Blue R concentrate is a methanol based stain especially designed for staining protein in polyacrylamide but will also stain agarose gels. The stain contains methanol and acetic acid so gels do not require fixing prior to staining.
Brilliant Blue R Concentrate has been used for protein band detection after nondenaturing gradient gel electrophoresis, isoelectric focussing and SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis).
For detection of protein bands following electrophoresis.

Reconstitution

After dilution with deionized water to a final volume of one liter, solution will contain 0.25% (w/v) Brilliant Blue R, 40% (v/v) methanol and 7% (v/v) acetic acid.

Legal Information

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization of soluble proteins in vapor diffusion for x-ray crystallography.
Benvenuti M and Mangani S
Nature Protocols, 2, 1633-1651 (2007)
Overexpression of human apolipoprotein AI in transgenic rats and the hyperlipoproteinemia associated with experimental nephrosis.
Burkey BF, et al.
Journal of Lipid Research, 36.7, 1463-1473 (1995)
The effect of ethanol or hepatotoxin exposure on rat transferrin desialylation.
Lof K, et al.
Alcohol and Alcoholism (Oxford, Oxfordshire), 31, 445-451 (1996)
Katharina Stock et al.
Cancer cell international, 19, 77-77 (2019-04-13)
The nucleation-promoting factor cortactin is expressed and promotes tumor progression and metastasis in various cancers. However, little is known about the biological role of cortactin in the progression of pancreatic ductal adenocarcinoma (PDAC). Cortactin and phosphorylated cortactin (Y421) were investigated
Melda Altikatoglu et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 39(3), 185-190 (2010-12-02)
Horseradish peroxidase (EC 1.11.1.7) was chemically modified by periodate-activated dextran. The activities of free and modified enzyme against organic-aqueous interface and some chemicals were determined. Modified HRP remained fully active in the presence of organic solvent for 4 h. However

Articles

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service