Merck
  • Simultaneous determination of bifonazole and tinctures of calendula flower in pharmaceutical creams by reversed-phase liquid chromatography.
Simultaneous determination of bifonazole and tinctures of calendula flower in pharmaceutical creams by reversed-phase liquid chromatography.
Journal of AOAC International (2006-03-11)
Carola F Ferreyra, Cristina S Ortiz
ABSTRACT
The aim of this research was to develop and validate a sensitive, rapid, easy, and precise reversed-phase liquid chromatography (LC) method for stability studies of bifonazole (I) formulated with tinctures of calendula flower (II). The method was especially developed for the analysis and quantitative determination of I and II in pure and combined forms in cream pharmaceutical formulations without using gradient elution and at room temperature. The influence on the stability of compound I of temperature, artificial radiation, and drug II used for the new pharmaceutical design was evaluated. The LC separation was carried out using a Supelcosil LC-18 column (25 cm x 4.6 mm id, 5 microm particle size); the mobile phase was composed of methanol-0.1 M ammonium acetate buffer (85 + 15, v/v) pumped isocratically at a flow rate of 1 mL/min; and ultraviolet detection was at 254 nm. The analysis time was less than 10 min. Calibration graphs were found to be linear in the 0.125-0.375 mg/mL (rI = 0.9991) and 0.639-1.916 mg/mL (rII = 0.9995) ranges for I and II, respectively. The linearity, precision, recovery, and limits of detection and quantification were satisfactory for I and II. The results obtained suggested that the developed LC method is selective and specific for the analysis of I and II in pharmaceutical products, and that it can be applied to stability studies.
Product Description
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material