Skip to Content
Merck
All Photos(1)

Documents

59613

Supelco

SUPELCOSIL LC-18 (5 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

Agency

suitable for USP L1

packaging

pkg of 2 ea

parameter

<70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix

silica gel high purity, spherical

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

compatibility

use to protect LC-18, LC-PAH

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Matar et al.
Therapeutic drug monitoring, 21(5), 559-566 (1999-10-16)
A simple, rapid, sensitive, and reproducible high-performance liquid chromatographic (HPLC) method for simultaneous determination of the antiepileptic drugs (ethosuximide, primidone, lamotrigine, phenobarbital, phenytoin, and carbamazepine) and two metabolites (carbamazepine-diol and carbamazepine-epoxide) in human plasma is described. The procedure involves extraction
O Cudina et al.
Farmaco (Societa chimica italiana : 1989), 55(2), 125-127 (2000-04-27)
Fluocortolone and its esters are synthetic corticosteroids used topically in the treatment of various skin disorders. A method that can be successfully used for the separation and determination of fluocortolone pivalate and fluocortolone hexanoate in suppositories was developed. This method
Carola F Ferreyra et al.
Journal of AOAC International, 88(6), 1649-1654 (2006-03-11)
The aim of this research was to develop and validate a sensitive, rapid, easy, and precise reversed-phase liquid chromatography (LC) method for stability studies of bifonazole (I) formulated with tinctures of calendula flower (II). The method was especially developed for
Mert Ilhan et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 578103-578103 (2014-08-29)
The aim of the present research is to evaluate the antiurolithiatic effect of the various extracts prepared from the fruits of Viburnum opulus L., in regard to its ethnobotanical record. To induce urolithiasis, 70 mg/kg sodium oxalate was injected to the
V Stocchi et al.
Analytical biochemistry, 146(1), 118-124 (1985-04-01)
A simple and rapid method for the determination of ATP, ADP, AMP, NADP+, NAD+, NADPH, and NADH in human erythrocytes is described. A single-step extraction procedure employing alkaline medium and CF 50A Amicon ultrafiltration membranes allows a simultaneous and total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service