Merck
  • Metabolic enzyme LDHA activates Rac1 GTPase as a noncanonical mechanism to promote cancer.
Metabolic enzyme LDHA activates Rac1 GTPase as a noncanonical mechanism to promote cancer.
Nature metabolism (2022-12-20)
Juan Liu, Cen Zhang, Tianliang Zhang, Chun-Yuan Chang, Jianming Wang, Ludvinna Bazile, Lanjing Zhang, Bruce G Haffty, Wenwei Hu, Zhaohui Feng
ABSTRACT
The glycolytic enzyme lactate dehydrogenase A (LDHA) is frequently overexpressed in cancer, which promotes glycolysis and cancer. The oncogenic effect of LDHA has been attributed to its glycolytic enzyme activity. Here we report an unexpected noncanonical oncogenic mechanism of LDHA; LDHA activates small GTPase Rac1 to promote cancer independently of its glycolytic enzyme activity. Mechanistically, LDHA interacts with the active form of Rac1, Rac1-GTP, to inhibit Rac1-GTP interaction with its negative regulator, GTPase-activating proteins, leading to Rac1 activation in cancer cells and mouse tissues. In clinical breast cancer specimens, LDHA overexpression is associated with higher Rac1 activity. Rac1 inhibition suppresses the oncogenic effect of LDHA. Combination inhibition of LDHA enzyme activity and Rac1 activity by small-molecule inhibitors displays a synergistic inhibitory effect on breast cancers with LDHA overexpression. These results reveal a critical oncogenic mechanism of LDHA and suggest a promising therapeutic strategy for breast cancers with LDHA overexpression.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-PAK1 (Ser199/Ser204) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-ARHGAP24 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TKS5 Antibody, clone 13H6.3, clone 13H6.3, from mouse
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.