Merck
  • Acetylation drives hepatocyte nuclear factor 1β stability by blocking proteasome-mediated degradation.
Acetylation drives hepatocyte nuclear factor 1β stability by blocking proteasome-mediated degradation.
Journal of cellular biochemistry (2018-12-14)
Filipa Lopes-Coelho, Fernanda Silva, Ana Hipólito, Bruno A Cardoso, Jacinta Serpa
ABSTRACT
Hepatocyte nuclear factor 1β (HNF1β) is mostly expressed in the liver, but is also expressed in other organs, like kidney, pancreas and genitourinary tract. In fact, HNF1β, a member of the superfamily of homeodomain-containing transcription factors, has been described as a hallmark in clear cell carcinomas. However, its role as an oncogene or as tumor suppressor gene remains controversial. Here, we disclose a mechanism of HNF1β stabilization and degradation, using human HNF1β-expressing cell lines of ovarian clear cell carcinoma (ES2), hepatocellular carcinoma (HEPG2), and normal immortalized kidney tubular cells (HK2). We show that increased levels of HNF1β is concomitant with an increase in the acetylation load and protein stabilization by interfering with the ubiquitin-proteasome degradation system. This study reinforces that acetylation, besides their role in regulating chromatin conformation and gene expression, could also act in the action, turnover and stability of proteins essential for the survival and progression of certain cancer types.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Anti-HNF1B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Anti-Acetyl Lysine Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-HNF1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial