Skip to Content
Merck
All Photos(9)

Key Documents

HPA002083

Sigma-Aldrich

Anti-HNF1B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

HNF1B Antibody - Anti-HNF1B antibody produced in rabbit, Hnf1B Antibody, Anti-HNF-1β, Anti-LFB3, Anti-MODY5, Anti-TCF2, Anti-VHNF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

KEVLVQALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDPWRAAKMIKGYMQQH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF1B(6928)

General description

HNF1B (hepatocyte nuclear factor 1 β), a homeodomain, is an atypical POU (Pit-1, Oct-1, andUnc-86) transcription factor which is associated with the early vertebrate development and embryonic survival. It is expressed in the pancreas, kidney, lung, ovary, testis, and throughout the gastrointestinal tract.

Immunogen

Hepatocyte nuclear factor 1β recombinant protein epitope signature tag (PrEST)

Application

Anti-HNF1B antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The expression of HNF1B (hepatocyte nuclear factor 1 β) is upregulated in papillary renal cell tumours (RCTs) as well as in some conventional RCCs, renal oncocytomas, chromophobe RCCs and normal kidneys. On the other hand, its expression is suppressed in Wilms′tumours. It is a nuclear protein required for liver-specific expression. It can binds to the promoters and enhancers of a variety of genes expressed in liver along with the proximal element (PE) of the albumin gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84742

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Laure Rougemont et al.
Human pathology, 81, 26-36 (2018-05-14)
Identification of the yolk sac tumor (YST) component in germ cell tumors (GCT) may prove challenging, and highly sensitive and specific immunohistochemical markers are still lacking. Preliminary data from the literature suggest that HNF1β may represent a sensitive marker of
James R Conner et al.
Cancer cytopathology, 123(5), 298-305 (2015-03-06)
Morphologic overlap between renal oncocytoma (RO) and chromophobe renal cell carcinoma (ChRCC) can pose diagnostic challenges, especially in biopsy samples in which tumor quantity may be limited, architectural features are not well represented, and gross examination is not possible. It
Martin Köbel et al.
The American journal of surgical pathology, 40(2), 166-180 (2015-10-23)
Mixed endometrial carcinoma refers to a tumor that comprises 2 or more distinct histotypes. We studied 18 mixed-type endometrial carcinomas-11 mixed serous and low-grade endometrioid carcinomas (SC/EC), 5 mixed clear cell and low-grade ECs (CCC/EC), and 2 mixed CCC and
Michaela Bártů et al.
Scientific reports, 10(1), 17151-17151 (2020-10-15)
Hepatocyte nuclear factor 1 beta (HNF1B) is a transcription factor which plays a crucial role in nephronogenesis, and its germline mutations have been associated with kidney developmental disorders. However, the effects of HNF1B somatic exonic mutations and its role in
Alicia A Goyeneche et al.
Cancer cell international, 15, 88-88 (2015-09-26)
We studied a primary culture developed from a biopsy of a clear cell carcinoma of the ovary (O-CCC) by (a) assessing its capacity to retain in vitro pathological features of the tumor of origin; (b) characterizing the main cells released

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service