Merck
  • Asparagine couples mitochondrial respiration to ATF4 activity and tumor growth.
Asparagine couples mitochondrial respiration to ATF4 activity and tumor growth.
Cell metabolism (2021-02-21)
Abigail S Krall, Peter J Mullen, Felicia Surjono, Milica Momcilovic, Ernst W Schmid, Christopher J Halbrook, Apisadaporn Thambundit, Steven D Mittelman, Costas A Lyssiotis, David B Shackelford, Simon R V Knott, Heather R Christofk
ABSTRACT
Mitochondrial respiration is critical for cell proliferation. In addition to producing ATP, respiration generates biosynthetic precursors, such as aspartate, an essential substrate for nucleotide synthesis. Here, we show that in addition to depleting intracellular aspartate, electron transport chain (ETC) inhibition depletes aspartate-derived asparagine, increases ATF4 levels, and impairs mTOR complex I (mTORC1) activity. Exogenous asparagine restores proliferation, ATF4 and mTORC1 activities, and mTORC1-dependent nucleotide synthesis in the context of ETC inhibition, suggesting that asparagine communicates active respiration to ATF4 and mTORC1. Finally, we show that combination of the ETC inhibitor metformin, which limits tumor asparagine synthesis, and either asparaginase or dietary asparagine restriction, which limit tumor asparagine consumption, effectively impairs tumor growth in multiple mouse models of cancer. Because environmental asparagine is sufficient to restore tumor growth in the context of respiration impairment, our findings suggest that asparagine synthesis is a fundamental purpose of tumor mitochondrial respiration, which can be harnessed for therapeutic benefit to cancer patients.
Sigma-Aldrich
L-Asparagine monohydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Rotenone, A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain.