Merck
  • Dissecting human embryonic skeletal stem cell ontogeny by single-cell transcriptomic and functional analyses.
Dissecting human embryonic skeletal stem cell ontogeny by single-cell transcriptomic and functional analyses.
Cell research (2021-01-22)
Jian He, Jing Yan, Jianfang Wang, Liangyu Zhao, Qian Xin, Yang Zeng, Yuxi Sun, Han Zhang, Zhijie Bai, Zongcheng Li, Yanli Ni, Yandong Gong, Yunqiao Li, Han He, Zhilei Bian, Yu Lan, Chunyu Ma, Lihong Bian, Heng Zhu, Bing Liu, Rui Yue
ABSTRACT
Human skeletal stem cells (SSCs) have been discovered in fetal and adult long bones. However, the spatiotemporal ontogeny of human embryonic SSCs during early skeletogenesis remains elusive. Here we map the transcriptional landscape of human limb buds and embryonic long bones at single-cell resolution to address this fundamental question. We found remarkable heterogeneity within human limb bud mesenchyme and epithelium, and aligned them along the proximal-distal and anterior-posterior axes using known marker genes. Osteo-chondrogenic progenitors first appeared in the core limb bud mesenchyme, which give rise to multiple populations of stem/progenitor cells in embryonic long bones undergoing endochondral ossification. Importantly, a perichondrial embryonic skeletal stem/progenitor cell (eSSPC) subset was identified, which could self-renew and generate the osteochondral lineage cells, but not adipocytes or hematopoietic stroma. eSSPCs are marked by the adhesion molecule CADM1 and highly enriched with FOXP1/2 transcriptional network. Interestingly, neural crest-derived cells with similar phenotypic markers and transcriptional networks were also found in the sagittal suture of human embryonic calvaria. Taken together, this study revealed the cellular heterogeneity and lineage hierarchy during human embryonic skeletogenesis, and identified distinct skeletal stem/progenitor cells that orchestrate endochondral and intramembranous ossification.
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-FoxP1 Antibody, serum, from rabbit