Merck
  • Isolation and Functional Identification of an Antiplatelet RGD-Containing Disintegrin from Cerastes cerastes Venom.
Isolation and Functional Identification of an Antiplatelet RGD-Containing Disintegrin from Cerastes cerastes Venom.
The protein journal (2020-09-23)
Meriem Ameziani, Fatah Chérifi, Hamida Kiheli, Samah Saoud, Ghania Hariti, Safia Kellou-Taîri, Fatima Laraba-Djebari
ABSTRACT
The current report focuses on purification, structural and functional characterization of Cerastategrin from Cerastes cerastes venom, a novel basic disintegrin (pI 8.36) with 128 amino acid residues and a molecular weight of 13 835.25 Da measured by MALDI-MSMS. The 3D structure of Cerastategrin is organized as α-helix (13%), β-strand (15%) and disordered structure (30%) and presents homologies with several snake venom disintegrins. Structural modeling shows that Cerastategrin presents an RGD motif that connects specifically to integrin receptors. Cerastategrin exhibits the inhibition of ADP induced platelets with an IC50 of 0.88 µg/mL and shows in vivo long stable anticoagulation effect 24 h post-injection of increasing doses ranging from 0.2 to 1 mg/kg, therefore, Cerastategrin maintained irreversibly the blood incoagulable. Moreover, Cerastategrin decreases the amount of bounded αIIbβ3 and reduced significantly the quantity of externalized P-Selectin. Cerastategrin acts as a molecule targeting specifically the receptor αIIbβ3; therefore, it behaves as a potent platelet activation inhibitor. As a new peptide with promising pharmacological properties, Cerastategrin could have a potential therapeutical effect in the vascular pathologies and may be a new effective treatment approach.
Sigma-Aldrich
Thrombin Receptor Activator Peptide 6, >95% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioXtra, 99.0-103.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Casein from bovine milk, technical grade
Sigma-Aldrich
[Glu1]-Fibrinopeptide B human, ≥90% (HPLC)
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sephadex® G-50, Superfine, Cytiva, pack of 100 g
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
1,10-Phenanthroline monohydrate, reagent grade