Merck
  • Convenient QSAR model for predicting the complexation of structurally diverse compounds with beta-cyclodextrins.
Convenient QSAR model for predicting the complexation of structurally diverse compounds with beta-cyclodextrins.
Bioorganic & medicinal chemistry (2008-12-06)
Alfonso Pérez-Garrido, Aliuska Morales Helguera, Adela Abellán Guillén, M Natália D S Cordeiro, Amalio Garrido Escudero
ABSTRACT
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation, and an efficient variable selection procedure, like the Genetic Algorithm, led to models with satisfactory global accuracy and predictivity. But the best-final QSAR model is based on Topological descriptors meanwhile offering a reasonable interpretation. This QSAR model was able to explain ca. 84% of the variance in the experimental activity, and displayed very good internal cross-validation statistics and predictivity on external data. It shows that the driving forces for CD complexation are mainly hydrophobic and steric (van der Waals) interactions. Thus, the results of our study provide a valuable tool for future screening and priority testing of beta-CDs guest molecules.
Product Description
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Supelco
Sulfisoxazole, VETRANAL®, analytical standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Menadione (K3), analytical standard
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Acridine, ≥96.5% (HPLC)
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
1,6-Hexanediol, 97%
Sigma-Aldrich
2-Pentanol, ≥98%, FG
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Methyl benzoate, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
3-Chlorophenol, 98%
Sigma-Aldrich
Isobutyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
3-Pentanol, 98%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%