Merck
Cerebellar 5HT-2A receptor mediates stress-induced onset of dystonia.
Science advances (2021-03-05)
Jung Eun Kim, Sujin Chae, Sungsoo Kim, Yeon-Joo Jung, Myoung-Goo Kang, Won Do Heo, Daesoo Kim
ABSTRACT
Stress is a key risk factor for dystonia, a debilitating motor disorder characterized by cocontractions of muscles leading to abnormal body posture. While the serotonin (5HT) system is known to control emotional responses to stress, its role in dystonia remains unclear. Here, we reveal that 5HT neurons in the dorsal raphe nuclei (DRN) send projections to the fastigial deep cerebellar nuclei (fDCN) and that photostimulation of 5HT-fDCN induces dystonia in wild-type mice. Moreover, we report that photoinhibition of 5HT-fDCN reduces dystonia in a1A tot/tot mice, a genetic model of stress-induced dystonia, and administration of a 5HT-2A receptor inverse agonist (MDL100907; 0.1 to 1 mg/kg) or shRNA-mediated knockdown of the ht2ar gene in fDCN can notably reduce the onset of dystonia in a1A tot/tot mice. These results support the serotonin theory of dystonia and suggest strategies for alleviating symptoms in human patients by blocking 5HT-2A receptors.
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Anti-Serotonin receptor 2A/HTR2A Antibody, clone 9B11.1, clone 9B11.1, from mouse
Sigma-Aldrich
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%