Merck
  • Tissue-Specific Regulation of the Wnt/β-Catenin Pathway by PAGE4 Inhibition of Tankyrase.
Tissue-Specific Regulation of the Wnt/β-Catenin Pathway by PAGE4 Inhibition of Tankyrase.
Cell reports (2020-07-23)
Sajjan Koirala, Jonathon Klein, Yumei Zheng, Nicole O Glenn, Travis Eisemann, Klementina Fon Tacer, Darcie J Miller, Ozlem Kulak, Meifen Lu, David B Finkelstein, Geoffrey Neale, Heather Tillman, Peter Vogel, Douglas W Strand, Lawrence Lum, Chad A Brautigam, John M Pascal, Wilson K Clements, Patrick Ryan Potts
ABSTRACT
Spatiotemporal control of Wnt/β-catenin signaling is critical for organism development and homeostasis. The poly-(ADP)-ribose polymerase Tankyrase (TNKS1) promotes Wnt/β-catenin signaling through PARylation-mediated degradation of AXIN1, a component of the β-catenin destruction complex. Although Wnt/β-catenin is a niche-restricted signaling program, tissue-specific factors that regulate TNKS1 are not known. Here, we report prostate-associated gene 4 (PAGE4) as a tissue-specific TNKS1 inhibitor that robustly represses canonical Wnt/β-catenin signaling in human cells, zebrafish, and mice. Structural and biochemical studies reveal that PAGE4 acts as an optimal substrate decoy that potently hijacks substrate binding sites on TNKS1 to prevent AXIN1 PARylation and degradation. Consistently, transgenic expression of PAGE4 in mice phenocopies TNKS1 knockout. Physiologically, PAGE4 is selectively expressed in stromal prostate fibroblasts and functions to establish a proper Wnt/β-catenin signaling niche through suppression of autocrine signaling. Our findings reveal a non-canonical mechanism for TNKS1 inhibition that functions to establish tissue-specific control of the Wnt/β-catenin pathway.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Glutathione Sepharose 4B, Cytiva 17-0756-05, pack of 100 mL
Sigma-Aldrich
Anti-poly-ADP-ribose binding reagent, Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse