Merck
  • Inhibitory effect of novel Eugenol Tosylate Congeners on pathogenicity of Candida albicans.
Inhibitory effect of novel Eugenol Tosylate Congeners on pathogenicity of Candida albicans.
BMC complementary medicine and therapies (2020-05-01)
Shabir Ahmad Lone, Aijaz Ahmad
ABSTRACT
The global prevalence of fungal diseases is increasing rapidly, which affects more than a billion people every year with significant mortality rate. On the other hand, the development of new drugs to treat these fungal infections is slow, while the current antifungal therapy is insufficient and associated with adverse side effects and emerging multidrug resistance. Therefore, development of novel antifungal drugs with least or no toxicity and multi-target mechanisms of action is an immediate priority. Natural products have long been known to possess antimicrobial activities and are source of new drugs. Currently, modifying natural products to synthesize derivatives/analogues are of great scientific focus for discovering novel drugs with improved potency and safety. Modifications in eugenol to synthesize eugenol derivatives with enhanced antifungal activity have already been reported. In this study, three most active novel eugenol tosylate congeners (ETC-5, ETC-6 and ETC-7) were selected from our previous study to investigate their effect on major virulence factors of Candida albicans which include adherence, morphogenesis, hydrolytic enzymes secretion, biofilm formation and on expression of genes related to these virulence factors. Adherence and biofilm formation were studied by alamarBlue dye and XTT reduction assays respectively, hydrolytic enzyme secretion was evaluated by plate assays. Further, morphological transition was monitored microscopically and RT-qPCR was used to assess the gene expression levels. ETCs significantly inhibited adherence in C. albicans with an inhibition range of 16-66%, and completely inhibited the morphogenesis at MIC values. Inhibition of proteinase and phospholipase activity was in the range of 2-48% and 8-34% respectively. Test compounds also significantly inhibit biofilm formation in C. albicans in the range of 7-77%. Furthermore, RT-qPCR results indicated a significant down regulation in expression levels of genes (ALS1, ALS2, ALS3, ALS9, CPH1, HWP1, SAP1, SAP2, SAP3 and PLB1) in C. albicans cells after treated with ETCs. The results indicated that these novel ETCs target major virulence factors of C. albicans and avert this commensal microbe to turn into pathogenic. However, further in-depth studies may facilitate the mechanisms involved by ETCs in targeting these virulence factors.
Product Description
Millipore
Peptone, mycological, suitable for microbiology
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Fetal Bovine Serum, γ-irradiated, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Calcium chloride, anhydrous, granular, ≤7.0 mm, ≥93.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
XTT sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%