Merck
  • PET, image-guided HDAC inhibition of pediatric diffuse midline glioma improves survival in murine models.
PET, image-guided HDAC inhibition of pediatric diffuse midline glioma improves survival in murine models.
Science advances (2020-08-25)
Umberto Tosi, Harikrishna Kommidi, Oluwaseyi Adeuyan, Hua Guo, Uday Bhanu Maachani, Nandi Chen, Taojunfeng Su, Guoan Zhang, David J Pisapia, Nadia Dahmane, Richard Ting, Mark M Souweidane
ABSTRACT
Efforts at altering the dismal prognosis of pediatric midline gliomas focus on direct delivery strategies like convection-enhanced delivery (CED), where a cannula is implanted into tumor. Successful CED treatments require confirmation of tumor coverage, dosimetry, and longitudinal in vivo pharmacokinetic monitoring. These properties would be best determined clinically with image-guided dosimetry using theranostic agents. In this study, we combine CED with novel, molecular-grade positron emission tomography (PET) imaging and show how PETobinostat, a novel PET-imageable HDAC inhibitor, is effective against DIPG models. PET data reveal that CED has significant mouse-to-mouse variability; imaging is used to modulate CED infusions to maximize tumor saturation. The use of PET-guided CED results in survival prolongation in mouse models; imaging shows the need of CED to achieve high brain concentrations. This work demonstrates how personalized image-guided drug delivery may be useful in potentiating CED-based treatment algorithms and supports a foundation for clinical translation of PETobinostat.
Product Description
Millipore
Phosphatase Inhibitor Cocktail Set V, 50X, Phosphatase Inhibitor Cocktail Set V is a 50X cocktail of four serine/threonine and tyrosine phosphatase inhibitors.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse