Merck
Dynamic Regulation of Mitochondrial Import by the Ubiquitin System.
Molecular cell (2020-03-07)
Lilian Phu, Christopher M Rose, Joy S Tea, Christopher E Wall, Erik Verschueren, Tommy K Cheung, Donald S Kirkpatrick, Baris Bingol
ABSTRACT
Mitochondria import nearly their entire proteome from the cytoplasm by translocating precursor proteins through the translocase of the outer membrane (TOM) complex. Here, we show dynamic regulation of mitochondrial import by the ubiquitin system. Acute pharmacological inhibition or genetic ablation of the mitochondrial deubiquitinase (DUB) USP30 triggers accumulation of Ub-substrates that are normally localized inside the mitochondria. Mitochondrial import of USP30 substrates is impaired in USP30 knockout (KO) cells, suggesting that deubiquitination promotes efficient import. Upstream of USP30, the E3 ligase March5 ubiquitinates mitochondrial proteins whose eventual import depends on USP30. In USP30 KOs, exogenous March5 expression induces accumulation of unimported translocation intermediates that are degraded by the proteasomes. In USP30 KO mice, TOM subunits have reduced abundance across multiple tissues. Together these data highlight how protein import into a subcellular compartment can be regulated by ubiquitination and deubiquitination by E3 ligase and DUB machinery positioned at the gate.
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-March5 Antibody, serum, from rabbit
Sigma-Aldrich
BAM15, ≥98% (HPLC)
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
Monoclonal Anti-MFN1 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution