Merck
  • Amino-modified Scholl-coupling mesoporous polymer for online solid-phase extraction of plant growth regulators from bean sprouts.
Amino-modified Scholl-coupling mesoporous polymer for online solid-phase extraction of plant growth regulators from bean sprouts.
Food chemistry (2020-04-03)
Shenghuai Hou, Xiaowei Sun, Lizong Chen, Yanzhen Yin, Wenhua Ji
ABSTRACT
A new amino-modified Scholl-coupling mesoporous polymer (NH2@SMPA)-online solid-phase extraction method, coupled with high-performance liquid chromatography (online SPE-HPLC) was established for the analysis of six plant growth regulators (PGRs) in bean sprouts. NH2@SMPA was synthesized by acid-catalyzed deacetylation of acetylamino-Scholl-coupling mesoporous polymer (SMPA). The diversity of functional groups, such as aromatic, acetylamino, and NH2, was conducive to multiple binding interactions between NH2@SMPA and PGRs. NH2@SMPA exhibited superior extraction capability for PGRs, compared with SMPA and commercial adsorbents. The extraction conditions, including loading solvent, pH of loading solution, eluting solvent, and flow rates of loading and elution, were optimized. Under the optimized conditions, wide linear ranges (0.01-500 μg kg-1) and low detection limits (2.34-20.2 ng kg-1) were obtained. The recoveries were satisfactory, i.e., 86.0% to 109% with relative standard deviations ≤9.8% (n = 3). Finally, the online SPE-HPLC method was successfully used for determination of PGRs in bean sprouts.
Sigma-Aldrich
4-Chlorophenoxyacetic acid, ≥98.0% (T)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%