Skip to Content
Merck
All Photos(3)

Documents

25890

Sigma-Aldrich

4-Chlorophenoxyacetic acid

≥98.0% (T)

Synonym(s):

4-CPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO3
CAS Number:
Molecular Weight:
186.59
Beilstein:
1211804
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

powder

SMILES string

OC(=O)COc1ccc(Cl)cc1

InChI

1S/C8H7ClO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

SODPIMGUZLOIPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorophenoxyacetic acid is a herbicide. Degradation of 4-chlorophenoxyacetic acid in aqueous medium by advanced electrochemical oxidation processes, using an undivided cell containing a Pt anode has been reported.

Application

  • Innovative adsorbents for chemical analysis: The application of three-dimensional material CZIF-8/CS-MS was explored as adsorbents for the determination of plant growth regulators including 4-Chlorophenoxyacetic acid in traditional Chinese medicine, improving the precision and efficiency of chemical residue analysis (Zhang et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sze Chieh Tan et al.
Mikrochimica acta, 188(2), 30-30 (2021-01-09)
A novel solvent-loaded dispersive solid-phase extraction (SL-DSPE) method integrated with liquid-phase microextraction (LPME) has been developed by the direct loading of solvent into the pores of a metal-organic framework (MOF), MIL-101(Cr)-NH2. Despite numerous advantages of MOFs, they are usually highly hydrophobic
Song Zhu et al.
Food science and biotechnology, 29(11), 1587-1595 (2020-10-23)
Determination of phytohormones have attracted increasing attentions in food safety field. In this study, an efficient and quantitative method was developed which can simultaneously determinate thirteen phytohormones in fruits and vegetables using solid phase extraction (SPE) combined with high performance
Sze Chieh Tan et al.
Talanta, 216, 120962-120962 (2020-05-28)
Two miniaturized sample preparation techniques, ultrasound-assisted emulsification microextraction (USAEME) and micro-solid-phase extraction (μ-SPE) have been integrated for the pre-concentration of four polar chlorophenoxy acid (CPA) herbicides from environmental aqueous samples. An metal-organic framework, MIL-101(Cr), characterized by its high porosity and
Shenghuai Hou et al.
Food chemistry, 321, 126702-126702 (2020-04-03)
A new amino-modified Scholl-coupling mesoporous polymer (NH2@SMPA)-online solid-phase extraction method, coupled with high-performance liquid chromatography (online SPE-HPLC) was established for the analysis of six plant growth regulators (PGRs) in bean sprouts. NH2@SMPA was synthesized by acid-catalyzed deacetylation of acetylamino-Scholl-coupling mesoporous
Ediz Yeşilkaya et al.
Hormone research, 72(4), 225-235 (2009-09-30)
Plant growth regulators are considered to leave minimal amounts of remnants and therefore cause no significant side effects in humans. In this study, we aimed to investigate the hormonal and histopathological effects of 4-chlorophenoxy acetic acid (4-CPA), a commonly used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service