Merck
  • Transcriptomics-Based Screening Identifies Pharmacological Inhibition of Hsp90 as a Means to Defer Aging.
Transcriptomics-Based Screening Identifies Pharmacological Inhibition of Hsp90 as a Means to Defer Aging.
Cell reports (2019-04-11)
Georges E Janssens, Xin-Xuan Lin, Lluís Millan-Ariño, Alan Kavšek, Ilke Sen, Renée I Seinstra, Nicholas Stroustrup, Ellen A A Nollen, Christian G Riedel
ABSTRACT
Aging strongly influences human morbidity and mortality. Thus, aging-preventive compounds could greatly improve our health and lifespan. Here we screened for such compounds, known as geroprotectors, employing the power of transcriptomics to predict biological age. Using age-stratified human tissue transcriptomes and machine learning, we generated age classifiers and applied these to transcriptomic changes induced by 1,309 different compounds in human cells, ranking these compounds by their ability to induce a "youthful" transcriptional state. Testing the top candidates in C. elegans, we identified two Hsp90 inhibitors, monorden and tanespimycin, which extended the animals' lifespan and improved their health. Hsp90 inhibition induces expression of heat shock proteins known to improve protein homeostasis. Consistently, monorden treatment improved the survival of C. elegans under proteotoxic stress, and its benefits depended on the cytosolic unfolded protein response-inducing transcription factor HSF-1. Taken together, our method represents an innovative geroprotector screening approach and was able to identify a class that acts by improving protein homeostasis.
Sigma-Aldrich
Prochlorperazine dimaleate salt
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sodium valproate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Radicicol from Diheterospora chlamydosporia, solid
Sigma-Aldrich
NU-1025, ≥97% (HPLC), solid
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Felbinac, European Pharmacopoeia (EP) Reference Standard
Santonin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimaprit dihydrochloride, ≥98% (NMR)
Sigma-Aldrich
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol, 98%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Trifluoperazine dihydrochloride, ≥99%, powder
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cantharidin
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)