Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-263-3
MDL number:
Product Name
Cantharidin,
Quality Segment
color
white
mp
215-217 °C (lit.), 215-218 °C
solubility
DMSO: soluble, ethanol: soluble
SMILES string
C[C@]12C3CCC(O3)[C@@]1(C)C(=O)OC2=O
InChI
1S/C10H12O4/c1-9-5-3-4-6(13-5)10(9,2)8(12)14-7(9)11/h5-6H,3-4H2,1-2H3/t5-,6+,9+,10-
InChI key
DHZBEENLJMYSHQ-XCVPVQRUSA-N
Gene Information
General description
Cantharidin is a poisonous vesicant, which is produced by blister bees. It is used to treat piles, ulcers, venomous worms and tuberculous scrofuloderma. It is used as an aphrodisiac, abortifacient and a veterinary medicine diuretic.
Application
Cantharidin has been used to inhibit protein phosphatase 2 (PP2A).
Biochem/physiol Actions
Inhibitor of protein phosphatase 2A.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Still not finding the right product?
Explore all of our products under Cantharidin
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C7632-25MG | 04061833520888 |
| C7632-100MG | 04061833520871 |
| C7632-500MG | 04061833629338 |
