Merck
  • Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734.
Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734.
Cancer research (2008-12-03)
Ana B Herrero, Alma M Astudillo, María A Balboa, Carmen Cuevas, Jesús Balsinde, Sergio Moreno
ABSTRACT
PM02734 is a novel synthetic antitumor drug that is currently in phase I clinical trials. To gain some insight into its mode of action, we used the yeast Saccharomyces cerevisiae as a model system. Treatment of S. cerevisiae with PM02734 rapidly induced necrosis-like cell death, as also found for mammalian cells treated with its close analogue kahalalide F. We have screened the complete set of 4,848 viable S. cerevisiae haploid deletion mutants to identify genes involved in sensitivity or resistance to PM02734. Forty-five percent of the 40 most sensitive strains identified had a role in intracellular vesicle trafficking, indicating that the drug severely affects this process. A mutant strain lacking the sphingolipid fatty acyl 2-hydroxylase Scs7 was found to be the most resistant to PM02734, whereas overexpression of Scs7 rendered the cells hypersensitive to PM02734. To validate these findings in human cells, we did small interfering RNA experiments and also overexpressed the Scs7 human homologue FA2H in human cancer cell lines. As in yeast, FA2H silencing turned the cells resistant to the drug, whereas FA2H overexpression led to an increased sensitivity. Moreover, exogenous addition of the 2-hydroxylated fatty acid 2-hydroxy palmitic acid to different human cell lines increased their sensitivity to the cytotoxic compound. Taken together, these results suggest that the cell membrane and, in particular, 2-hydroxy fatty acid-containing ceramides are important for PM02734 activity. These findings may have important implications in the development of PM02734 because tumor cells with high FA2H expression are expected to be particularly sensitive to this drug.
Product Description
Avanti
18:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860830P, powder
Avanti
16:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860815P, powder
Avanti
20:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860820P, powder
Avanti
24:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860824P, powder
Avanti
24:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860825P, powder
Avanti
12:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860811P, powder
Avanti
18:1(2S-OH) Ceramide, Avanti Research - A Croda Brand 860828P, powder
Avanti
20:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860819P, powder
Avanti
22:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860822P, powder
Avanti
18:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860829P, powder
Avanti
24:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860823P, powder
Avanti
18:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860827P, powder
Avanti
16:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860816P, powder
Avanti
22:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860821P, powder
Avanti
12:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860812P, powder
Avanti
17:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860817P, powder
Avanti
17:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860818P, powder