Skip to Content
Merck
All Photos(1)

Key Documents

860820P

Avanti

20:0(2S-OH) Ceramide

Avanti Research - A Croda Brand 860820P, powder

Synonym(s):

N-(2′-(S)-hydroxyarachidoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H75NO4
CAS Number:
Molecular Weight:
610.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860820P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860820P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C38H75NO4/c1-3-5-7-9-11-13-15-17-18-19-21-23-25-27-29-31-33-37(42)38(43)39-35(34-40)36(41)32-30-28-26-24-22-20-16-14-12-10-8-6-4-2/h30,32,35-37,40-42H,3-29,31,33-34H2,1-2H3,(H,39,43)/b32-30+/t35-,36+,37-/m0/s1

InChI key

CFYLSUZUMMPFNC-GBPFXZCASA-N

General description

Ceramides linked to 2-hydroxy fatty acids (hFA) are present in the surface epithelium of the skin. 20:0(2S-OH) Ceramide is a unique ceramide containing 20C long chain base fatty acid (arachidic acid)-with 2′-hydroxyl group in S configuration.

Biochem/physiol Actions

Hydroxy fatty acid (hFA)-sphingolipids help in formation and function of myelin. In addition, they also play a vital role in cell signaling, cell differentiation and apoptosis. In epidermis, hFA-ceramides aid in permeability barrier function. NAD(P)H-dependent enzyme, fatty acid 2-hydroxylase (FA2H) catalyzes the synthesis of hFA-ceramides. FA2H gene mutation leads to the development of neurological disorders such as leukodystrophy and spastic paraparesis in humans. hFA-ceramides help PM02734 (elisidepsin), an antitumor drug to exhibit its activity.

Packaging

5 mL Amber Glass Screw Cap Vial (860820P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PM02734 (elisidepsin) induces caspase-independent cell death associated with features of autophagy, inhibition of the Akt/mTOR signaling pathway, and activation of death-associated protein kinase
Ling YH, et al.
Clinical Cancer Research, 17(16), 5353-5366 (2011)
Yi-He Ling et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(16), 5353-5366 (2011-06-22)
PM02734 (elisidepsin) is a synthetic marine-derived cyclic peptide of the kahalalide family currently in phase II clinical development. The mechanisms of cell death induced by PM02734 remain unknown. Human non-small-cell lung cancer (NSCLC) cell lines H322 and A549 were used
Stereospecificity of fatty acid 2-hydroxylase and differential functions of 2-hydroxy fatty acid enantiomers
Guo L, et al.
Journal of Lipid Research, 53(7), 1327-1335 (2012)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Helena Maier et al.
The Journal of biological chemistry, 286(29), 25922-25934 (2011-06-02)
2-Hydroxylated fatty acid (HFA)-containing sphingolipids are abundant in mammalian skin and are believed to play a role in the formation of the epidermal barrier. Fatty acid 2-hydroxylase (FA2H), required for the synthesis of 2-hydroxylated sphingolipids in various organs, is highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service