Merck
  • Diastereomer-specific quantification of bioactive hexosylceramides from bacteria and mammals.
Diastereomer-specific quantification of bioactive hexosylceramides from bacteria and mammals.
Journal of lipid research (2017-04-05)
Johanna von Gerichten, Kerstin Schlosser, Dominic Lamprecht, Ivan Morace, Matthias Eckhardt, Dagmar Wachten, Richard Jennemann, Hermann-Josef Gröne, Matthias Mack, Roger Sandhoff
ABSTRACT
Mammals synthesize, cell-type specifically, the diastereomeric hexosylceramides, β-galactosylceramide (GalCer) and β-glucosylceramide (GlcCer), which are involved in several diseases, such as sphingolipidosis, diabetes, chronic kidney diseases, or cancer. In contrast, Bacteroides fragilis, a member of the human gut microbiome, and the marine sponge, Agelas mauritianus, produce α-GalCer, one of the most potent stimulators for invariant natural killer T cells. To dissect the contribution of these individual stereoisomers to pathologies, we established a novel hydrophilic interaction chromatography-based LC-MS2 method and separated (R > 1.5) corresponding diastereomers from each other, independent of their lipid anchors. Testing various bacterial and mammalian samples, we could separate, identify (including the lipid anchor composition), and quantify endogenous β-GlcCer, β-GalCer, and α-GalCer isomers without additional derivatization steps. Thereby, we show a selective decrease of β-GlcCers versus β-GalCers in cell-specific models of GlcCer synthase-deficiency and an increase of specific β-GlcCers due to loss of β-glucoceramidase 2 activity. Vice versa, β-GalCer increased specifically when cerebroside sulfotransferase (Gal3st1) was deleted. We further confirm β-GalCer as substrate of globotriaosylceramide synthase for galabiaosylceramide synthesis and identify additional members of the human gut microbiome to contain immunogenic α-GalCers. Finally, this method is shown to separate corresponding hexosylsphingosine standards, promoting its applicability in further investigations.
Product Description
Avanti
C16 Galactosyl(β) Ceramide (d18:1/16:0), Avanti Research - A Croda Brand 860521P, powder
Avanti
C18(2S-OH) Galactosyl(β) Ceramide, Avanti Research - A Croda Brand
Avanti
C24:1 Galactosyl(α) Ceramide (d18:1/24:1(15Z)), D-galactosyl- α-1,1′ N-nervonoyl- D-erythro-sphingosine, powder
Avanti
C16 Glucosyl(β) Ceramide (d18:1/16:0), Avanti Research - A Croda Brand
Avanti
C24:1 Galactosyl(β) Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand 860546P, powder
Avanti
C18(2R-OH) Galactosyl(β) Ceramide, Avanti Research - A Croda Brand 860840P, powder
Avanti
C16 Galactosyl(α) Ceramide (d18:1/16:0), D-galactosyl-α-1,1′ N-palmitoyl- D-erythro-sphingosine, powder
Avanti
C24:1 Glucosyl(β) Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand 860549P, powder