Skip to Content
Merck
All Photos(1)

Key Documents

860539P

Avanti

C16 Glucosyl(β) Ceramide (d18:1/16:0)

Avanti Research - A Croda Brand

Synonym(s):

D-glucosyl-β-1,1′-N-palmitoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H77NO8
CAS Number:
Molecular Weight:
700.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (860539P-25mg)
pkg of 1 × 5 mg (860539P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

InChI key

VJLLLMIZEJJZTE-NNTBDIJYSA-N

General description

Glucosyl(β) Ceramide (β-GlcCer) is a naturally occurring glycolipid and an ubiquitous intracellular metabolite. It might be present in the endoplasmic reticulum or Golgi apparatus.

Application

C16 Glucosyl(β) Ceramide (d18:1/16:0) or D-glucosyl-β-1,1′-N-palmitoyl-D-erythro-sphingosine has been used as a stock solution for the in vitro treatment of bacteria and to study the role of sphingolipids on pulmonary Pseudomonas aeruginosa infections in cystic fibrosis (CF). It may be used in the quantification of lipid in green fluorescence protein (GFP) and fatty acid transport protein 4 (FATP4) cells using electrospray ionization tandem mass spectrometry (ESI-MS/MS).

Biochem/physiol Actions

Glucosyl(β) Ceramide (β-GlcCer) is an endogenous macrophage inducible C-type lectin (Mincle) ligand, which has immunostimulatory property. It also stimulates inflammatory and acquired immune responses on myeloid cells through Mincle. It is a potential biomarker in Gaucher′s disease. β-GlcCer regulates the function of natural killer T (NKT) lymphocytes and exhibits antimetastatic effects. A balanced level of intracellular β-GlcCer is essential for maintaining tissue homeostasis.

Packaging

5 mL Amber Glass Screw Cap Vial (860539P-25mg)
5 mL Amber Glass Screw Cap Vial (860539P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Barbara Kovacic et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 41(3), 1208-1218 (2017-04-21)
Cystic fibrosis (CF) is the most common autosomal-recessive disorder in western countries. Previous studies have demonstrated an important role of sphingolipids in the pathophysiology of cystic fibrosis. It has been shown that ceramide has a central role in various pulmonary
Masashi Inafuku et al.
Lipids, 47(6), 581-591 (2012-03-20)
Natural killer (NK) T cells are well known to play important roles in both tumor rejection and the defense against infectious. Therefore, the antitumor potential of NKT cell-activating antigens have been the focus for the development of NKT cell-based immunotherapies.
Marko Jović et al.
Molecular biology of the cell, 23(8), 1533-1545 (2012-02-18)
Gaucher disease is a lysosomal storage disorder caused by a defect in the degradation of glucosylceramide catalyzed by the lysosomal enzyme β-glucocerebrosidase (GBA). GBA reaches lysosomes via association with its receptor, lysosomal integral membrane protein type 2 (LIMP-2). We found
Masahiro Nagata et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(16), E3285-E3294 (2017-04-05)
Sensing and reacting to tissue damage is a fundamental function of immune systems. Macrophage inducible C-type lectin (Mincle) is an activating C-type lectin receptor that senses damaged cells. Notably, Mincle also recognizes glycolipid ligands on pathogens. To elucidate endogenous glycolipids
E Zigmond et al.
Gut, 56(1), 82-89 (2006-12-19)
beta-Glucosylceramide, a naturally occurring glycolipid, exerts modulatory effects on natural killer T (NKT) lymphocytes. To determine whether beta-glucosylceramide can alter NKT function in opposite directions, colitis was induced by intracolonic installation of trinitrobenzenesulphonic acid, and hepatocellular carcinoma (HCC) was induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service