Merck
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology.
Biochimica et biophysica acta (2009-12-23)
Hiroko Hama
ABSTRACT
2-Hydroxy fatty acids (hFA) are important components of a subset of mammalian sphingolipids. The presence of hFA in sphingolipids is best described in the nervous system, epidermis, and kidney. However, the literature also indicates that various hFA-sphingolipids are present in additional tissues and cell types, as well as in tumors. Biosynthesis of hFA-sphingolipids requires fatty acid 2-hydroyxlase, and degradation of hFA-sphingolipids depends, at least in part, on lysosomal acid ceramidase and the peroxisomal fatty acid alpha-oxidation pathway. Mutations in the fatty acid 2-hydroxylase gene, FA2H, have been associated with leukodystrophy and spastic paraparesis in humans, underscoring the importance of hFA-sphingolipids in the nervous system. In the epidermis, hFA-ceramides are essential for the permeability barrier function. Physiological function of hFA-sphingolipids in other organs remains largely unknown. Recent evidence indicates that hFA-sphingolipids have specific roles in cell signaling.
Product Description
Avanti
18:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860830P, powder
Avanti
16:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860815P, powder
Avanti
20:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860820P, powder
Avanti
24:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860824P, powder
Avanti
24:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860825P, powder
Avanti
12:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860811P, powder
Avanti
18:1(2S-OH) Ceramide, Avanti Research - A Croda Brand 860828P, powder
Avanti
20:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860819P, powder
Avanti
18:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860827P, powder
Avanti
16:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860816P, powder
Avanti
22:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860821P, powder
Avanti
12:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860812P, powder
Avanti
17:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860817P, powder
Avanti
17:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860818P, powder
Avanti
22:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860822P, powder
Avanti
18:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860829P, powder
Avanti
24:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860823P, powder