Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon dioxide
Synonym(s):
Sand, white quartz, Quartz, Silicon dioxide
Linear Formula:
SiO
2
CAS Number:
14808-60-7
Molecular Weight:
60.08
EC Number:
238-878-4
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Particle Size (µm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S5631
Silicon dioxide,
~99%, 0.5-10 μm (approx. 80% between 1-5 μm)
Expand
View Pricing
31624
Silicon dioxide,
acid washed and calcined, Analytical Reagent
Expand
View Pricing
31623
Silicon dioxide,
washed and calcined, analytical reagent
Expand
View Pricing